3-Fluoro-2-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A141559-250MG A141559-1G A141559-25G A141559-10G A141559-100MG A141559-5G A141559-5G
77027-578EA 141.11 USD
77027-578 77316-178 77027-576 103803-012 77316-176 77027-580 103834-046
3-Fluoro-2-nitrobenzoic acid 98%
3-Fluoro-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368342
CAS Number: 1000339-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR