3-Fluoro-2-methylphenol 98%

Supplier: Ambeed
Warning

A851761-1G A851761-25G A851761-5G A851761-100G A851761-10G A851761-250MG
103841-242EA 22 USD
103841-242 76845-496 76845-498 76845-500 76981-802 76981-804 76981-806
3-Fluoro-2-methylphenol 98%
3-Fluoro-2-methylphenol

Precaución: For research use only

Formula: C₇H₇FO
Boiling Pt: 187.9 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD03788546
CAS Number: 443-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR