3-Fluoro-o-anisidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Fluoro-2-methoxyaniline, 3-Fluoro-2-anisidine, 2-Amino-6-fluoroanisole
A194892-1G
A194892-25G
A194892-10G
A194892-250MG
A194892-100G
103816-702EA
8
USD
103816-702
103847-316
77085-254
77085-256
77164-386
77164-388
77892-259
3-Fluoro-o-anisidine 97%
3-Fluoro-o-anisidine
Precaución: for Research Use Only
Formula:
C₇H₈FNO Boiling Pt: 217.9 °C (760 mm Hg) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007759 CAS Number: 437-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...