3-Fluoro-2-iodobenzonitrile 97%

Supplier: Ambeed
Warning

A205474-100MG A205474-1G A205474-5G A205474-250MG
103817-632EA 33 USD
103817-632 103848-298 76770-304 77086-512 77086-514 77877-637
3-Fluoro-2-iodobenzonitrile 97%
3-Fluoro-2-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
Melting Pt: 88…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07782075
CAS Number: 916792-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR