3-Fluoro-2-iodoaniline hydrochloride 97%

Supplier: Ambeed
Warning

A170414-100MG A170414-10G A170414-250MG A170414-1G A170414-5G
103806-486EA 23 USD
103806-486 103844-762 76973-976 76973-978 76973-980 77081-874 77081-876
3-Fluoro-2-iodoaniline hydrochloride 97%
3-Fluoro-2-iodoaniline hydrochloride

Precaución: for Research Use Only

Formula: C₆H₆ClFIN
MW: 273.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19982814
CAS Number: 122455-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR