3-Fluoro-2-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoropyridine-2-carboxaldehyde, 3-Fluoropicolinaldehyde, 3-Fluoropyridine-2-carbaldehyde, 3-Fluoro-2-pyridinecarboxaldehyde

A280401-100G A280401-10G A280401-250MG A280401-1G A280401-5G A280401-25G
103813-506EA 11 USD
103813-506 103853-452 77002-136 77002-138 77002-140 77093-386 77093-388 77305-356
3-Fluoro-2-formylpyridine 98%
3-Fluoro-2-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄FNO
Melting Pt: 49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781234
CAS Number: 31224-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR