3-Fluoro-2,2-dimethylpropanoic acid 97%

Supplier: Ambeed
Danger

A308671-100MG A308671-250MG A308671-1G
103814-568EA 57 USD
103814-568 103835-744 76816-870 76816-872 76838-502
3-Fluoro-2,2-dimethylpropanoic acid 97%
3-Fluoro-2,2-dimethylpropanoic acid

Precaución: for Research Use Only

Formula: C5H9FO2
Storage Temperature: Ambient
MDL Number: MFCD17677039
CAS Number: 64241-77-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR