3-Ethynylthiophene 95%

Supplier: Ambeed
Danger

A691488-1G A691488-25G A691488-5G A691488-10G
103840-584EA 60 USD
103840-584 76844-800 76844-802 76844-804 77280-762
3-Ethynylthiophene 95%
3-Ethynylthiophene

Precaución: for Research Use Only

Formula: C₆H₄S
Boiling Pt: 152…153 °C (760 mm Hg)
Flash Pt: 46.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039973
CAS Number: 67237-53-0
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR