3-Ethynylpyridine 97%

Supplier: Ambeed
Danger

Synonyms: 3-Pyridylacetylene

A655024-1G A655024-25G A655024-5G A655024-10G A655024-250MG
103840-412EA 38 USD
103840-412 76844-598 76844-600 76844-602 77280-542 77280-544
3-Ethynylpyridine 97%
3-Ethynylpyridine

Precaución: for Research Use Only

Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 170.5 °C (760 mm Hg)
Melting Pt: 39…40 °C
Flash Pt: 56.6 °C
Storage Temperature: Freezer
MDL Number: MFCD02177459
CAS Number: 2510-23-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR