3-Ethynylbenzaldehyde 97%

Supplier: Ambeed
Warning

A117387-100MG A117387-1G
103798-874EA 68 USD
103798-874 103824-758 77257-980 77297-538
3-Ethynylbenzaldehyde 97%
3-Ethynylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆O
MW: 130.1433 g/mol
Melting Pt: 75…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06245349
CAS Number: 77123-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR