3-Ethynylanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethynyl-3-methoxybenzene, (3-Methoxyphenyl)ethyne, (3-Methoxyphenyl)acetylene, 1-Methoxy-3-ethynylbenzene

A178019-1G A178019-5G A178019-100G A178019-10G
103807-292EA 12 USD
103807-292 103845-632 77083-030 77083-032 77273-934 77273-936
3-Ethynylanisole 97%
3-Ethynylanisole

Precaución: for Research Use Only

Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 204.4 °C (760 mm Hg)
Flash Pt: 76.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00160810
CAS Number: 768-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR