3-Ethynylaniline hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A256066-100G
A256066-10G
A256066-5G
A256066-1G
A256066-25G
103812-236EA
5
USD
103812-236
103852-096
76988-040
76988-042
77091-572
77304-666
77304-668
3-Ethynylaniline hydrochloride 95%
3-Ethynylaniline hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₈ClN Boiling Pt: 275.5 °C (760 mm Hg) Melting Pt: 193…197 °C Flash Pt: 120.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12913913 CAS Number: 207226-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...