3-Ethoxypyridin-4-amine 97%

Supplier: Ambeed
Warning

A158068-250MG A158068-1G A158068-5G A158068-100MG
103805-088EA 230 USD
103805-088 103843-296 76824-016 77319-140 77319-142 77885-280
3-Ethoxypyridin-4-amine 97%
3-Ethoxypyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD16877218
CAS Number: 89943-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR