3-(Ethoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

A141248-100G A141248-10G A141248-1G A141248-5G
103802-968EA 5 USD
103802-968 77162-322 77162-324 77316-112 77316-114
3-(Ethoxycarbonyl)phenylboronic acid 98%
3-(Ethoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
Boiling Pt: 368.4 °C (760 mm Hg)
Melting Pt: 131…136 °C
Flash Pt: 176.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02179444
CAS Number: 4334-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR