3-Ethoxyacrylic acid 98%

Supplier: Ambeed
Warning

A131443-100G A131443-10G A131443-25G A131443-1G A131443-5G
103801-582EA 5 USD
103801-582 77272-012 77272-014 77272-016 77298-484 77314-086
3-Ethoxyacrylic acid 98%
3-Ethoxyacrylic acid

Precaución: for Research Use Only

Formula: C5H8O3
Boiling Pt: 109 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01566841
CAS Number: 6192-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR