3-Cyclohexylpropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyclohexylpropanoic acid, 3-Cyclohexylpropionic Acid

A149772-100G A149772-10G A149772-1000G A149772-25G A149772-5G
103808-730EA 135 USD
103808-730 103842-204 77272-940 77272-942 77317-710 77317-712 77317-714
3-Cyclohexylpropionic acid 98%
3-Cyclohexylpropionic acid

Precaución: for Research Use Only

Formula: C₉H₁₆O₂
MW: 156.22 g/mol
Boiling Pt: 276.5 °C (760 mm Hg)
Melting Pt: 14…17 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001527
CAS Number: 701-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR