3-Cyanophenylboronic acid 98%

Supplier: Ambeed
Warning

A156495-100G A156495-10G A156495-500G A156495-25G A156495-5G
103804-896EA 9 USD
103804-896 103843-096 77137-960 77137-962 77137-964 77318-882 77318-884
3-Cyanophenylboronic acid 98%
3-Cyanophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BNO₂
Boiling Pt: 347.4 °C (760 mm Hg)
Melting Pt: 298 °C
Flash Pt: 163.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01318967
CAS Number: 150255-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR