3-Cyanobenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Formylbenzonitrile

A386419-25G A386419-5G A386419-10G A386419-100G A386419-500G
103837-836EA 5 USD
103837-836 76841-192 76841-194 77003-704 77568-786 77672-914
3-Cyanobenzaldehyde 98%
3-Cyanobenzaldehyde

Precaución: For research use only.

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 210.8 °C (760 mm Hg)
Melting Pt: 75…78 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR