3-Cyano-4-hydroxybenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A231253-1G
A231253-250MG
103810-766EA
79
USD
103810-766
103850-514
77089-396
77275-756
3-Cyano-4-hydroxybenzoic acid 95%
3-Cyano-4-hydroxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₅NO₃ Boiling Pt: 392.447 °C (760 mm Hg) Flash Pt: 191.145 °C Storage Temperature: Ambient |
MDL Number:
MFCD09998350 CAS Number: 70829-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...