3-Cyano-4-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A241772-1G A241772-100MG A241772-250MG
103811-428EA 25 USD
103811-428 103851-234 77090-396 77276-026 77276-028
3-Cyano-4-fluorobenzoic acid 97%
3-Cyano-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C8H4FNO2
MW: 165.12 g/mol
Boiling Pt: 322.4 °C (760 mm Hg)
Flash Pt: 106.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03094339
CAS Number: 171050-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR