3-Cyano-4-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-4-fluorobenzeneboronic acid

A237024-100G A237024-100MG A237024-10G A237024-250MG A237024-1G A237024-25G A237024-5G
103811-124EA 23 USD
103811-124 103850-900 77000-324 77000-326 77000-328 77000-330 77089-960 77089-962 77089-964
3-Cyano-4-fluorophenylboronic acid 98%
3-Cyano-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
Boiling Pt: 333.2 °C (760 mm Hg)
Melting Pt: >300 °C
Flash Pt: 155.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03095130
CAS Number: 214210-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR