3-Cyano-2-methylphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A134022-10G
A134022-250MG
A134022-25G
A134022-1G
A134022-5G
103801-954EA
123
USD
103801-954
77095-106
77095-108
77298-678
77314-664
77314-666
3-Cyano-2-methylphenylboronic acid 97%
3-Cyano-2-methylphenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₈BNO₂ MW: 160.966 g/mol Boiling Pt: 359.8±52.0 °C (760 mm Hg) Flash Pt: 171.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18394417 CAS Number: 313546-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...