3-Cyano-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Indazole-3-carbonitrile

A237331-1G A237331-5G A237331-10G A237331-250MG
103811-148EA 55 USD
103811-148 103850-926 77090-006 77090-008 77165-684 77165-686
3-Cyano-1H-indazole 95%
3-Cyano-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₅N₃
MW: 143.15 g/mol
Boiling Pt: 370.3 °C (760 mm Hg)
Flash Pt: 126.669 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01860240
CAS Number: 50264-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR