3-Chloro-2-thenoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-thiophenecarboxylic acid, 3-Chlorothiophene-2-carboxylic acid

A330588-1G A330588-100G A330588-10G A330588-250MG A330588-25G
103825-556EA 10 USD
103825-556 103836-518 76839-500 77290-422 77290-424 77290-426 77306-410
3-Chloro-2-thenoic acid 98%
3-Chloro-2-thenoic acid

Precaución: for Research Use Only

Formula: C₅H₃ClO₂S
Melting Pt: 186…190 °C
Flash Pt: 130.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00043888
CAS Number: 59337-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR