3-Chloroquinoline 98%

Supplier: Ambeed
Warning

A168743-1G A168743-25G A168743-5G A168743-100G A168743-100MG A168743-250MG A168743-10G
103806-314EA 33 USD
103806-314 103844-568 77081-628 77081-630 77081-632 77273-624 77273-626 77273-628 77748-124
3-Chloroquinoline 98%
3-Chloroquinoline

Precaución: for Research Use Only

Formula: C9H6ClN
MW: 163.6036 g/mol
Boiling Pt: 255.7 °C (760 mm Hg)
Flash Pt: 133.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00234489
CAS Number: 612-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR