3-Chloro-2-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloropyridine-2-carboxaldehyde

A117449-250MG A117449-25G A117449-1G A117449-5G
103798-890EA 37 USD
103798-890 103824-774 76781-484 76781-486 77311-558 77311-560
3-Chloro-2-formylpyridine 97%
3-Chloro-2-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Boiling Pt: 201.5 °C (760 mm Hg)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09991855
CAS Number: 206181-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR