3-Chloropyrazine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A227138-100G
A227138-10G
A227138-250MG
A227138-1G
A227138-25G
A227138-5G
103819-300EA
20
USD
103819-300
103850-206
76987-196
76987-198
76987-200
77088-966
77303-790
77303-792
3-Chloropyrazine-2-carboxylic acid 95%
3-Chloropyrazine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C5H3ClN2O2 Boiling Pt: 317 °C (760 mm Hg) Flash Pt: 145.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318438 CAS Number: 27398-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...