2-Amino-3-chloropyrazine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloropyrazin-2-amine, 3-Chloro-2-pyrazinamine
A200064-25G
A200064-5G
A200064-100G
A200064-10G
103847-788EA
8
USD
103847-788
77085-846
77085-848
77164-534
77164-536
2-Amino-3-chloropyrazine 98%
2-Amino-3-chloropyrazine
Precaución: for Research Use Only
Formula:
C₄H₄ClN₃ MW: 129.55 g/mol Boiling Pt: 248.1 °C (760 mm Hg) Melting Pt: 167…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114305 CAS Number: 6863-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...