3-Chlorophenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane
A103947-100G
A103947-10G
A103947-25G
A103947-500G
A103947-5G
103822-336EA
9
USD
103822-336
77283-168
77283-170
77283-172
77283-174
77309-202
3-Chlorophenylboronic acid 98%
3-Chlorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₆BClO₂ MW: 156.38 g/mol Boiling Pt: 311.4 °C (760 mm Hg) Melting Pt: 185…189 °C Flash Pt: 142.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161354 CAS Number: 63503-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...