N-Methyl-3-chloropropylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Methylamino)propyl Chloride Hydrochloride, 3-Chloro-N-methylpropan-1-amine hydrochloride

A162006-100G A162006-1G A162006-5G A162006-10G
103805-560EA 6 USD
103805-560 103843-806 76720-152 77319-830 77319-832 77667-266
N-Methyl-3-chloropropylamine hydrochloride 95%
N-Methyl-3-chloropropylamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClN
MW: 144.04 g/mol
Boiling Pt: 180.8 °C (760 mm Hg)
Flash Pt: 63.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00144935
CAS Number: 97145-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR