3-Chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-N-methyl-5-(trifluoromethyl)pyridin-2-amine

A637716-1G A637716-25G A637716-5G
103840-322EA 69 USD
103840-322 76844-494 76844-496 76844-498
3-Chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine 98%
3-Chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine

Precaución: for Research Use Only

Formula: C₇H₆ClF₃N₂
MW: 210.59 g/mol
Boiling Pt: 216.3 °C (760 mm Hg)
Flash Pt: 84.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06375956
CAS Number: 89810-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR