3-Chloro-N,N-dimethylpropan-1-amine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Dimethylaminopropyl chloride

A211398-10G A211398-500G A211398-1G A211398-25G A211398-5G A211398-100G
103818-108EA 8 USD
103818-108 103848-802 76999-620 76999-622 77303-196 77303-198 77303-200 77593-384
3-Chloro-N,N-dimethylpropan-1-amine 98%
3-Chloro-N,N-dimethylpropan-1-amine

Precaución: for Research Use Only

Formula: C5H12ClN
Boiling Pt: 130.7 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00044496
CAS Number: 109-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR