3-Chloroisonicotinic acid 97%

Supplier: Ambeed
Warning

A237176-25G A237176-5G A237176-1G A237176-10G
103811-134EA 5 USD
103811-134 103850-912 77089-982 77089-984 77304-076 77883-626
3-Chloroisonicotinic acid 97%
3-Chloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
Melting Pt: 227…231 °C
Flash Pt: 187.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03094685
CAS Number: 88912-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR