3-Chloro-4-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloroisonicotinaldehyde, 3-Chloro-4-pyridinecarboxaldehyde, 3-Chloropyridine-4-carboxaldehyde

A169273-10G A169273-250MG A169273-1G
103806-368EA 7 USD
103806-368 103844-622 76813-154 76813-156 77081-698
3-Chloro-4-formylpyridine 97%
3-Chloro-4-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 58…62 °C
Flash Pt: 89.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06200712
CAS Number: 72990-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR