3-Chloro-6-methylpyridazine 98%

Supplier: Ambeed
Warning

A334396-25G A334396-5G A334396-100G A334396-10G A334396-1G
103836-624EA 14 USD
103836-624 76839-632 76839-634 77015-462 77015-464 77015-466
3-Chloro-6-methylpyridazine 98%
3-Chloro-6-methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 58…62 °C
Flash Pt: 134.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052905
CAS Number: 1121-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR