3-Chloro-6-methoxypyridazine 97%

Supplier: Ambeed
Warning

A257524-100G A257524-10G A257524-25G A257524-5G
103852-182EA 9 USD
103852-182 77014-106 77014-108 77091-692 77091-694
3-Chloro-6-methoxypyridazine 97%
3-Chloro-6-methoxypyridazine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
Boiling Pt: 284.9 °C (760 mm Hg)
Melting Pt: 84…85 °C
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00006467
CAS Number: 1722-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR