3-Chloro-6-hydrazinopyridazine 98%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-6-hydrazinylpyridazine

A294202-1G A294202-25G A294202-5G A294202-10G
103814-002EA 5 USD
103814-002 103835-142 76837-770 76837-772 76837-774 76875-532
3-Chloro-6-hydrazinopyridazine 98%
3-Chloro-6-hydrazinopyridazine

Precaución: for Research Use Only

Formula: C₄H₅ClN₄
MW: 144.56 g/mol
Boiling Pt: 402.2 °C (760 mm Hg)
Melting Pt: 137…141 °C
Flash Pt: 197 °C
Storage Temperature: Freezer
MDL Number: MFCD00051740
CAS Number: 17284-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR