3-Chloro-6-fluorobenzo[d]isoxazole 97%

Supplier: Ambeed
Warning

A197772-100MG A197772-250MG
103816-966EA 204 USD
103816-966 103847-588 76975-122 76975-124
3-Chloro-6-fluorobenzo[d]isoxazole 97%
3-Chloro-6-fluorobenzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₇H₃ClFNO
MW: 171.556 g/mol
Boiling Pt: 255.4±20.0 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD16996105
CAS Number: 374554-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR