3-Chloro-6-(chloromethyl)pyridazine 95%

Supplier: Ambeed
Danger

A881618-100MG A881618-250MG A881618-1G
103841-310EA 525 USD
103841-310 76820-852 76820-854 76845-580
3-Chloro-6-(chloromethyl)pyridazine 95%
3-Chloro-6-(chloromethyl)pyridazine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD10698627
CAS Number: 120276-59-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR