3-Chloro-5-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-chloro-α,α,α-trifluortoluene, 5-Chloro-α,α,α-trifluoro-m-toluidine, 3-Amino-5-chlorobenzotrifluoride, 3-Amino-5-chloro-α,α,α-trifluorotoluene, 3-Chloro-5-trifluoromethylaniline

A135464-10G A135464-1G A135464-25G A135464-5G
103802-160EA 6 USD
103802-160 76811-486 77314-990 77314-992 77314-994
3-Chloro-5-(trifluoromethyl)aniline 98%
3-Chloro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
MW: 195.57
Flash Pt: 85.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02684210
CAS Number: 69411-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR