3-Chloro-5-nitroaniline 98%

Supplier: Ambeed
Warning

A270285-100MG A270285-10G A270285-250MG A270285-1G
103812-996EA 16 USD
103812-996 103852-902 76875-046 76875-048 76875-050 77305-052
3-Chloro-5-nitroaniline 98%
3-Chloro-5-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
Boiling Pt: 332.7 °C (760 mm Hg)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD01009865
CAS Number: 5344-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR