3-Chloro-5-methylpyridine 98%

Supplier: Ambeed
Warning

A311849-1G A311849-5G A311849-25G
103814-682EA 11 USD
103814-682 103835-872 76838-682 76838-684 77306-060
3-Chloro-5-methylpyridine 98%
3-Chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 176.7 °C (760 mm Hg)
Flash Pt: 75.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04114217
CAS Number: 19230-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR