3-Chloro-5-methylaniline 98%

Supplier: Ambeed
Warning

A100372-25G A100372-5G A100372-10G
103795-398EA 6 USD
103795-398 103821-098 77025-128 77296-654 77592-056
3-Chloro-5-methylaniline 98%
3-Chloro-5-methylaniline

Precaución: for Research Use Only

Formula: C7H8ClN
MW: 141.60 g/mol
Storage Temperature: Freezer
MDL Number: MFCD15527197
CAS Number: 29027-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR