3-Chloro-5-methoxyphenol 97%

Supplier: Ambeed
Warning

A258565-1G A258565-5G A258565-10G A258565-250MG
103812-386EA 17 USD
103812-386 103852-262 77091-794 77091-796 77276-488 77276-490
3-Chloro-5-methoxyphenol 97%
3-Chloro-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MW: 158.582 g/mol
Boiling Pt: 265.3 °C (760 mm Hg)
Melting Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002260
CAS Number: 65262-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR