3-Chloro-5-methoxybenzoic acid 97%
Supplier: Ambeed
|
Danger
|
A184751-1G
A184751-25G
A184751-5G
A184751-10G
A184751-250MG
103815-778EA
14
USD
103815-778
103846-344
77083-960
77083-962
77083-964
77164-004
77164-006
3-Chloro-5-methoxybenzoic acid 97%
3-Chloro-5-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇ClO₃ Melting Pt: 172…176 °C Flash Pt: 149.227 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01317994 CAS Number: 82477-67-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...