3-Chloro-5-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A258067-1G A258067-5G A258067-10G
103812-358EA 7 USD
103812-358 103852-230 77091-758 77091-760 77748-484
3-Chloro-5-hydroxybenzonitrile 98%
3-Chloro-5-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD11226540
CAS Number: 473923-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR