3-Chloro-5-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A125212-250MG A125212-25G A125212-1G A125212-5G A125212-10G
103800-642EA 20 USD
103800-642 77161-566 77161-568 77312-788 77312-790 77683-113
3-Chloro-5-hydroxybenzoic acid 95%
3-Chloro-5-hydroxybenzoic acid

Precaución: For research use only.

Formula: C₇H₅ClO₃
Boiling Pt: 364.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04114327
CAS Number: 53984-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR