3-Chloro-5H-pyrrolo[2,3-b]pyrazine 97%

Supplier: Ambeed
Warning

A138359-100MG A138359-250MG A138359-1G
103802-606EA 66 USD
103802-606 76972-454 76972-456 77298-988
3-Chloro-5H-pyrrolo[2,3-b]pyrazine 97%
3-Chloro-5H-pyrrolo[2,3-b]pyrazine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.569 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12024539
CAS Number: 1111638-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR