3-Chloro-5-fluoroaniline 97%

Supplier: Ambeed
Warning

A373276-25G A373276-5G A373276-100G A373276-10G A373276-500G A373276-1G
103826-530EA 9 USD
103826-530 103837-548 76840-812 76840-814 77290-888 77290-890 77290-892 77306-852
3-Chloro-5-fluoroaniline 97%
3-Chloro-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 227.8 °C (760 mm Hg)
Flash Pt: 91.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03407962
CAS Number: 4863-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR