3-Chloro-5-cyanobenzoic acid 95%

Supplier: Ambeed
Warning

A279796-100MG A279796-250MG
103813-484EA 246 USD
103813-484 76977-380 76977-382
3-Chloro-5-cyanobenzoic acid 95%
3-Chloro-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂
MW: 181.58 g/mol
Boiling Pt: 329.0±27.0 °C (760 mm Hg)
Flash Pt: 152.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06797228
CAS Number: 327056-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR